Merck
  • Cholinergic axons in the rat ventral tegmental area synapse preferentially onto mesoaccumbens dopamine neurons.
Cholinergic axons in the rat ventral tegmental area synapse preferentially onto mesoaccumbens dopamine neurons.
The Journal of comparative neurology (2005-12-31)
Natalia Omelchenko, Susan R Sesack
ABSTRACT
Cholinergic afferents to the ventral tegmental area (VTA) contribute substantially to the regulation of motivated behaviors and the rewarding properties of nicotine. These actions are believed to involve connections with dopamine (DA) neurons projecting to the nucleus accumbens (NAc). However, this direct synaptic link has never been investigated, nor is it known whether cholinergic inputs innervate other populations of DA and gamma-aminobutyric acid (GABA) neurons, including those projecting to the prefrontal cortex (PFC). We addressed these questions by using electron microscopic analysis of retrograde tract-tracing and immunocytochemistry for the vesicular acetylcholine transporter (VAChT) and for tyrosine hydroxylase (TH) and GABA. In tissue labeled for TH, VAChT(+) terminals frequently synapsed onto DA mesoaccumbens neurons but only seldom contacted DA mesoprefrontal cells. In tissue labeled for GABA, one-third of VAChT(+) terminals innervated GABA-labeled dendrites, including both mesoaccumbens and mesoprefrontal populations. VAChT(+) synapses onto DA and mesoaccumbens neurons were more commonly of the asymmetric (presumed excitatory) morphological type, whereas VAChT(+) synapses onto GABA cells were more frequently symmetric (presumed inhibitory or modulatory). These findings suggest that cholinergic inputs to the VTA mediate complex synaptic actions, with a major portion of this effect likely to involve an excitatory influence on DA mesoaccumbens neurons. As such, the results suggest that natural and drug rewards operating through cholinergic afferents to the VTA have a direct synaptic link to the mesoaccumbens DA neurons that modulate approach behaviors.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Pricing and availability is not currently available.