Merck
  • Capture of microRNA-bound mRNAs identifies the tumor suppressor miR-34a as a regulator of growth factor signaling.
Capture of microRNA-bound mRNAs identifies the tumor suppressor miR-34a as a regulator of growth factor signaling.
PLoS genetics (2011-11-22)
Ashish Lal, Marshall P Thomas, Gabriel Altschuler, Francisco Navarro, Elizabeth O'Day, Xiao Ling Li, Carla Concepcion, Yoon-Chi Han, Jerome Thiery, Danielle K Rajani, Aaron Deutsch, Oliver Hofmann, Andrea Ventura, Winston Hide, Judy Lieberman
ABSTRACT
A simple biochemical method to isolate mRNAs pulled down with a transfected, biotinylated microRNA was used to identify direct target genes of miR-34a, a tumor suppressor gene. The method reidentified most of the known miR-34a regulated genes expressed in K562 and HCT116 cancer cell lines. Transcripts for 982 genes were enriched in the pull-down with miR-34a in both cell lines. Despite this large number, validation experiments suggested that ~90% of the genes identified in both cell lines can be directly regulated by miR-34a. Thus miR-34a is capable of regulating hundreds of genes. The transcripts pulled down with miR-34a were highly enriched for their roles in growth factor signaling and cell cycle progression. These genes form a dense network of interacting gene products that regulate multiple signal transduction pathways that orchestrate the proliferative response to external growth stimuli. Multiple candidate miR-34a-regulated genes participate in RAS-RAF-MAPK signaling. Ectopic miR-34a expression reduced basal ERK and AKT phosphorylation and enhanced sensitivity to serum growth factor withdrawal, while cells genetically deficient in miR-34a were less sensitive. Fourteen new direct targets of miR-34a were experimentally validated, including genes that participate in growth factor signaling (ARAF and PIK3R2) as well as genes that regulate cell cycle progression at various phases of the cell cycle (cyclins D3 and G2, MCM2 and MCM5, PLK1 and SMAD4). Thus miR-34a tempers the proliferative and pro-survival effect of growth factor stimulation by interfering with growth factor signal transduction and downstream pathways required for cell division.
Sigma-Aldrich
Anti-CCNG2 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-AGBL5 (AB1) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-PSMD5 antibody produced in mouse, clone 3E2, purified immunoglobulin, buffered aqueous solution