Merck
  • A general protocol for the generation of Nanobodies for structural biology.
A general protocol for the generation of Nanobodies for structural biology.
Nature protocols (2014-03-01)
Els Pardon, Toon Laeremans, Sarah Triest, Søren G F Rasmussen, Alexandre Wohlkönig, Armin Ruf, Serge Muyldermans, Wim G J Hol, Brian K Kobilka, Jan Steyaert
ABSTRACT
There is growing interest in using antibodies as auxiliary tools to crystallize proteins. Here we describe a general protocol for the generation of Nanobodies to be used as crystallization chaperones for the structural investigation of diverse conformational states of flexible (membrane) proteins and complexes thereof. Our technology has a competitive advantage over other recombinant crystallization chaperones in that we fully exploit the natural humoral response against native antigens. Accordingly, we provide detailed protocols for the immunization with native proteins and for the selection by phage display of in vivo-matured Nanobodies that bind conformational epitopes of functional proteins. Three representative examples illustrate that the outlined procedures are robust, making it possible to solve by Nanobody-assisted X-ray crystallography in a time span of 6-12 months.
Sigma-Aldrich
Methyltrioctylammonium hydrogen sulfate, ≥95.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG