Merck
  • Proteomic and immunochemical characterization of glutathione transferase as a new allergen of the nematode Ascaris lumbricoides.
Proteomic and immunochemical characterization of glutathione transferase as a new allergen of the nematode Ascaris lumbricoides.
PloS one (2013-11-14)
Nathalie Acevedo, Jens Mohr, Josefina Zakzuk, Martin Samonig, Peter Briza, Anja Erler, Anna Pomés, Christian G Huber, Fatima Ferreira, Luis Caraballo
ABSTRACT
Helminth infections and allergy have evolutionary and clinical links. Infection with the nematode Ascaris lumbricoides induces IgE against several molecules including invertebrate pan-allergens. These antibodies influence the pathogenesis and diagnosis of allergy; therefore, studying parasitic and non-parasitic allergens is essential to understand both helminth immunity and allergy. Glutathione transferases (GSTs) from cockroach and house dust mites are clinically relevant allergens and comparative studies between them and the GST from A. lumbricoides (GSTA) are necessary to evaluate their allergenicity. We sought to analyze the allergenic potential of GSTA in connection with the IgE response to non-parasitic GSTs. IgE to purified GSTs from Ascaris (nGSTA and rGSTA), house dust mites (rDer p 8, nBlo t 8 and rBlo t 8), and cockroach (rBla g 5) was measured by ELISA in subjects from Cartagena, Colombia. Also, multidimensional proteomic approaches were used to study the extract of A. lumbricoides and investigate the existence of GST isoforms. We found that among asthmatics, the strength of IgE levels to GSTA was significantly higher than to mite and cockroach GSTs, and there was a strong positive correlation between IgE levels to these molecules. Specific IgE to GSTA was found in 13.2% of controls and 19.5% of asthmatics. In addition nGSTA induced wheal and flare in skin of sensitized asthmatics indicating that it might be of clinical relevance for some patients. Frequency and IgE levels to GSTA were higher in childhood and declined with age. At least six GST isoforms in A. lumbricoides bind human IgE. Four isoforms were the most abundant and several amino acid substitutions were found, mainly on the N-terminal domain. In conclusion, a new allergenic component of Ascaris has been discovered; it could have clinical impact in allergic patients and influence the diagnosis of mite and cockroach allergy in tropical environments.
Sigma-Aldrich
Anti-Human IgE−Alkaline Phosphatase antibody, Mouse monoclonal, clone GE-1, purified from hybridoma cell culture
Pricing and availability is not currently available.