Merck
  • Supramolecular Self-Assemblies with Nanoscale RGD Clusters Promote Cell Growth and Intracellular Drug Delivery.
Supramolecular Self-Assemblies with Nanoscale RGD Clusters Promote Cell Growth and Intracellular Drug Delivery.
ACS applied materials & interfaces (2016-10-21)
Fengyang Xu, Jie Liu, Jian Tian, Linfeng Gao, Xiaju Cheng, Yue Pan, Ziling Sun, Xinming Li
ABSTRACT
In this work, we reported the generation of a novel supramolecular hydrogelator from a peptide derivative which consisted of a structural motif (e.g., Fc-FF) for supramolecular self-assembly and a functional moiety (e.g., RGD) for integrin binding. Following self-assembly in water at neutral pH, this molecule first tended to form metastable spherical aggregates, which subsequently underwent a morphological transformation to form high-aspect-ratio nanostructures over 2 h when aged at room temperature. More importantly, because of the presence of nanoscale RGD clusters on the surface of nanostructures, the self-assembled nanomaterials (e.g., nanoparticles and nanofibers) can be potentially used as a biomimetic matrix for cell culture and as a vector for cell-targeting drug delivery via multivalent RGD-integrin interactions.
Sigma-Aldrich
Fmoc-Asp(OtBu)-OH, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Pricing and availability is not currently available.