Merck
  • A Novel Selective Inhibitor of Delta-5 Desaturase Lowers Insulin Resistance and Reduces Body Weight in Diet-Induced Obese C57BL/6J Mice.
A Novel Selective Inhibitor of Delta-5 Desaturase Lowers Insulin Resistance and Reduces Body Weight in Diet-Induced Obese C57BL/6J Mice.
PloS one (2016-11-11)
Hiroaki Yashiro, Shuichi Takagahara, Yumiko Okano Tamura, Ikuo Miyahisa, Junji Matsui, Hideo Suzuki, Shota Ikeda, Masanori Watanabe
ABSTRACT
Obesity is now recognized as a state of chronic low-grade inflammation and is called as metabolic inflammation. Delta-5 desaturase (D5D) is an enzyme that metabolizes dihomo-γ-linolenic acid (DGLA) to arachidonic acid (AA). Thus, D5D inhibition increases DGLA (precursor to anti-inflammatory eicosanoids) while decreasing AA (precursor to pro-inflammatory eicosanoids), and could result in synergistic improvement in the low-grade inflammatory state. Here, we demonstrate reduced insulin resistance and the anti-obesity effect of a D5D selective inhibitor (compound-326), an orally active small-molecule, in a high-fat diet-induced obese (DIO) mouse model. In vivo D5D inhibition was confirmed by determining changes in blood AA/DGLA profiles. In DIO mice, chronic treatment with compound-326 lowered insulin resistance and caused body weight loss without significant impact on cumulative calorie intake. Decreased macrophage infiltration into adipose tissue was expected from mRNA analysis. Increased daily energy expenditure was also observed following administration of compound-326, in line with sustained body weight loss. These data indicate that the novel D5D selective inhibitor, compound-326, will be a new class of drug for the treatment of obese and diabetic patients.
Sigma-Aldrich
Sibutramine hydrochloride monohydrate, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
D5D-IN-326, ≥98% (HPLC)
Pricing and availability is not currently available.