Merck
  • Pharmacological targeting of CXCL12/CXCR4 signaling in prostate cancer bone metastasis.
Pharmacological targeting of CXCL12/CXCR4 signaling in prostate cancer bone metastasis.
Molecular cancer (2016-11-05)
M Katie Conley-LaComb, Louie Semaan, Rajareddy Singareddy, Yanfeng Li, Elisabeth I Heath, Seongho Kim, Michael L Cher, Sreenivasa R Chinni
ABSTRACT
The CXCL12/CXCR4 axis transactivates HER2 and promotes intraosseous tumor growth. To further explore the transactivation of HER2 by CXCL12, we investigated the role of small GTP protein G We used a variety of methods such as lipid raft isolation, invasion assays, an in vivo model of intratibial tumor growth, bone histomorphometry, and immunohistochemistry to determine the role of CXCR4 signaling in lipid raft membrane microdomains and effects of targeting of CXCR4 for bone tumor growth. We determined that (a) CXCL12/CXCR4 transactivation of EGFR and HER2 is confined to lipid raft membrane microdomains, (b) CXCL12 activation of HER2 and Src is mediated by small GTP proteins in lipid rafts, (c) inhibition of the CXCL12/CXCR4 axis through plerixafor abrogates the initial establishment of tumor growth without affecting the growth of established bone tumors, and (d) inhibition of EGFR signaling through gefitinib leads to inhibition of established bone tumor growth. These data suggest that lipid raft membrane microdomains are key sites for CXCL12/CXCR4 transactivation of HER2 via small GTP binding protein G
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Src