- Chromosome dynamics: new light on Aurora B kinase function.
Chromosome dynamics: new light on Aurora B kinase function.
Current biology : CB (2002-07-18)
Katie B Shannon, E D Salmon
PMID12121637
ABSTRACT
Aurora B family kinases play an essential role in chromosome segregation and cytokinesis. Recent work suggests that the kinase activity is required for bipolar chromosome orientation, kinetochore assembly, spindle checkpoint and microtubule dynamics. Aurora B also has additional functions in chromosome condensation and cohesion.
Sigma-Aldrich
AURORA B, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution