Merck
  • A regulatory loop between miR-132 and miR-125b involved in gonadotrope cells desensitization to GnRH.
A regulatory loop between miR-132 and miR-125b involved in gonadotrope cells desensitization to GnRH.
Scientific reports (2016-08-20)
Jérôme Lannes, David L'hôte, Ambra Fernandez-Vega, Ghislaine Garrel, Jean-Noël Laverrière, Joëlle Cohen-Tannoudji, Bruno Quérat
ABSTRACT
The GnRH neurohormone is the main activator of the pituitary gonadotropins, LH and FSH. Here we investigated the contribution of microRNAs in mediating GnRH activation. We first established that miR-125b targets several actors of Gαq/11 signalling pathway, without altering Gαs pathway. We then showed that a Gαs-mediated, PKA-dependent phosphorylation of NSun2 methyltransferase leads to miR-125b methylation and thereby induces its down-regulation. We demonstrated that NSun2 mRNA is a target of miR-132 and that NSun2 may be inactivated by the PP1α phosphatase. Time-course analysis of GnRH treatment revealed an initial NSun2-dependent down-regulation of miR-125b with consecutive up-regulation of LH and FSH expression. Increase of miR-132 and of the catalytic subunit of PP1α then contributed to NSun2 inactivation and to the return of miR-125b to its steady-state level. The Gαq/11-dependent pathway was thus again silenced, provoking the down-regulation of LH, FSH and miR-132. Overall, this study reveals that a regulatory loop that tends to maintain or restore high and low levels of miR-125b and miR-132, respectively, is responsible for gonadotrope cells desensitization to sustained GnRH. A dysregulation of this loop might be responsible for the inverted dynamics of these two miRNAs reported in several neuronal and non-neuronal pathologies.
Sigma-Aldrich
Anti-N6-methyladenosine (m6A) Antibody, from rabbit
Sigma-Aldrich
Anti-Gq/11α Antibody, CT, Upstate®, from rabbit