Merck
  • An expeditious synthesis of tamoxifen, a representative SERM (selective estrogen receptor modulator), via the three-component coupling reaction among aromatic aldehyde, cinnamyltrimethylsilane, and beta-chlorophenetole.
An expeditious synthesis of tamoxifen, a representative SERM (selective estrogen receptor modulator), via the three-component coupling reaction among aromatic aldehyde, cinnamyltrimethylsilane, and beta-chlorophenetole.
Bioorganic & medicinal chemistry (2007-10-02)
Isamu Shiina, Yoshiyuki Sano, Kenya Nakata, Masahiko Suzuki, Toshikazu Yokoyama, Akane Sasaki, Tomoko Orikasa, Tomomi Miyamoto, Masahiko Ikekita, Yukitoshi Nagahara, Yoshimune Hasome
ABSTRACT
Two new synthetic pathways to the anti-cancer agent tamoxifen and its derivatives were developed. The first route involved the aldol reaction of benzyl phenyl ketone with acetaldehyde followed by Friedel-Crafts substitution with anisole in the presence of Cl(2)Si(OTf)(2) to produce 1,1,2-triaryl-3-acetoxybutane, a precursor of the tamoxifen derivatives. The second one utilized the novel three-component coupling reaction among aromatic aldehydes, cinnamyltrimethylsilane, and aromatic nucleophiles using HfCl(4) as a Lewis acid catalyst to produce 3,4,4-triarylbutene, that is also a valuable intermediate of the tamoxifen derivatives. The former strategy requires a total of 10 steps from the aldol formation to the final conversion to tamoxifen, whereas the latter needs only three or four steps to produce tamoxifen and droloxifene including the installation of the side-chain moiety and the base-induced double-bond migration to form the tetra-substituted olefin structure. This synthetic strategy seems to serve as a new and practical pathway to prepare not only the tamoxifen derivatives but also the other SERMs (selective estrogen receptor modulators) including estrogen-dependent breast cancer and osteoporosis agents.
Sigma-Aldrich
Ethoxybenzene, 99%
Sigma-Aldrich
Tamoxifen, ≥99%