- Organocatalysis with a fluorous tag: asymmetric reduction of imines with trichlorosilane catalyzed by amino acid-derived formamides.
Organocatalysis with a fluorous tag: asymmetric reduction of imines with trichlorosilane catalyzed by amino acid-derived formamides.
The Journal of organic chemistry (2007-02-10)
Andrei V Malkov, Marek Figlus, Sigitas Stoncius, Pavel Kocovský
PMID17288379
ABSTRACT
Asymmetric reduction of ketimines 1 with trichlorosilane can be catalyzed by N-methylvaline-derived Lewis-basic formamides 3a-d with high enantioselectivity (< or =95% ee) and low catalyst loading (1-5 mol %) at room temperature in toluene. Appending a fluorous tag, as in 5a-c, simplifies the isolation procedure, while preserving high enantioselectivity (< or =92% ee).