Merck
  • Discovery and SAR of biaryl piperidine MCH1 receptor antagonists through solid-phase encoded combinatorial synthesis.
Discovery and SAR of biaryl piperidine MCH1 receptor antagonists through solid-phase encoded combinatorial synthesis.
Bioorganic & medicinal chemistry letters (2005-06-28)
Tao Guo, Yuefei Shao, Gang Qian, Laura L Rokosz, Tara M Stauffer, Rachael C Hunter, Suresh D Babu, Huizhong Gu, Doug W Hobbs
ABSTRACT
An encoded combinatorial library based on aryl and biaryl piperidine scaffolds was designed and synthesized. Screening of this library resulted in the discovery of high-nanomolar biaryl piperidine-based MCH1 receptor antagonists. Follow-up optimization using a parallel synthesis provided potent, single digit nanomolar antagonists.
Sigma-Aldrich
Bromocyclobutane, 96%
Pricing and availability is not currently available.