Merck
Catalytic enantioselective allylation of ketoimines.
Journal of the American Chemical Society (2006-06-08)
Reiko Wada, Tomoyuki Shibuguchi, Sae Makino, Kounosuke Oisaki, Motomu Kanai, Masakatsu Shibasaki
ABSTRACT
A general catalytic allylation of simple ketoimines was developed using 1 mol % of CuF.3PPh(3) as catalyst, 1.5 mol % of La(O(i)Pr)(3) as the cocatalyst, and stable and nontoxic allylboronic acid pinacol ester as the nucleophile. This reaction constituted a good template for developing the first catalytic enantioselective allylation of ketoimines. In this case, using LiO(i)Pr as the cocatalyst produced higher enantioselectivity and reactivity than La(O(i)Pr)(3). Thus, using the CuF-cyclopentyl-DuPHOS complex (10 mol %) and LiO(i)Pr (30 mol %) in the presence of (t)BuOH (1 equiv) produced high enantioselectivity up to 93% ee from a range of aromatic ketoimines. Mechanistic studies indicated that LiO(i)Pr accelerates the reaction by increasing the concentration of an active nucleophile, allylcopper.
Sigma-Aldrich
Lanthanum(III) isopropoxide, 98%
Pricing and availability is not currently available.