Merck
  • Repeated low-dose kainate administration in C57BL/6J mice produces temporal lobe epilepsy pathology but infrequent spontaneous seizures.
Repeated low-dose kainate administration in C57BL/6J mice produces temporal lobe epilepsy pathology but infrequent spontaneous seizures.
Experimental neurology (2016-02-21)
Anthony D Umpierre, Isaiah V Bennett, Lismore D Nebeker, Thomas G Newell, Bruce B Tian, Kyle E Thomson, H Steve White, John A White, Karen S Wilcox
ABSTRACT
More efficient or translationally relevant approaches are needed to model acquired temporal lobe epilepsy (TLE) in genetically tractable mice. The high costs associated with breeding and maintaining transgenic, knock-in, or knock-out lines place a high value on the efficiency of induction and animal survivability. Herein, we describe our approaches to model acquired epilepsy in C57BL/6J mice using repeated, low-dose kainate (KA) administration paradigms. Four paradigms (i.p.) were tested for their ability to induce status epilepticus (SE), temporal lobe pathology, and the development of epilepsy. All four paradigms reliably induce behavioral and/or electrographic SE without mortality over a 7d period. Two of the four paradigms investigated produce features indicative of TLE pathology, including hippocampal cell death, widespread astrogliosis, and astrocyte expression of mGluR5, a feature commonly reported in TLE models. Three of the investigated paradigms were able to produce aberrant electrographic features, such as interictal spiking in cortex. However, only one paradigm, previously published by others, produces spontaneous recurrent seizures over an eight week period. Presentation of spontaneous seizures is rare (N=2/14), with epilepsy preferentially developing in animals having a high number of seizures during SE. Overall, repeated, low-dose KA administration improves the efficiency and pathological relevance of a systemic KA insult, but does not produce a robust epilepsy phenotype under the experimental paradigms described herein.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Metabotropic Glutamate Receptor 5 Antibody, pain, Chemicon®, from rabbit
Pricing and availability is not currently available.