Merck
Enantioselective organocatalytic reductive amination.
Journal of the American Chemical Society (2006-01-05)
R Ian Storer, Diane E Carrera, Yike Ni, David W C MacMillan
ABSTRACT
The first enantioselective organocatalytic reductive amination reaction has been accomplished. The development of a new chiral phosphoric acid catalyst has provided a convenient strategy for the enantioselective construction of protected primary amines and provided a highly stereoselective method for the reductive amination of heterocyclic amines. A diverse spectrum of ketone and amine substrates can be accommodated in high yield and excellent enantioselectivity. This new protocol realizes a key benefit of reductive amination versus imine reduction, in that ketimines derived from alkyl-alkyl ketones are unstable to isolation, a fundamental limitation that is comprehensively bypassed using this direct organocatalytic reductive amination.
Sigma-Aldrich
(R)-(–)-3,3′-Bis(triphenylsilyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate, 95%