Merck
  • VEGF-A isoforms program differential VEGFR2 signal transduction, trafficking and proteolysis.
VEGF-A isoforms program differential VEGFR2 signal transduction, trafficking and proteolysis.
Biology open (2016-04-06)
Gareth W Fearnley, Gina A Smith, Izma Abdul-Zani, Nadira Yuldasheva, Nadeem A Mughal, Shervanthi Homer-Vanniasinkam, Mark T Kearney, Ian C Zachary, Darren C Tomlinson, Michael A Harrison, Stephen B Wheatcroft, Sreenivasan Ponnambalam
ABSTRACT
Vascular endothelial growth factor A (VEGF-A) binding to the receptor tyrosine kinase VEGFR2 triggers multiple signal transduction pathways, which regulate endothelial cell responses that control vascular development. Multiple isoforms of VEGF-A can elicit differential signal transduction and endothelial responses. However, it is unclear how such cellular responses are controlled by isoform-specific VEGF-A-VEGFR2 complexes. Increasingly, there is the realization that the membrane trafficking of receptor-ligand complexes influences signal transduction and protein turnover. By building on these concepts, our study shows for the first time that three different VEGF-A isoforms (VEGF-A165, VEGF-A121 and VEGF-A145) promote distinct patterns of VEGFR2 endocytosis for delivery into early endosomes. This differential VEGFR2 endocytosis and trafficking is linked to VEGF-A isoform-specific signal transduction events. Disruption of clathrin-dependent endocytosis blocked VEGF-A isoform-specific VEGFR2 activation, signal transduction and caused substantial depletion in membrane-bound VEGFR1 and VEGFR2 levels. Furthermore, such VEGF-A isoforms promoted differential patterns of VEGFR2 ubiquitylation, proteolysis and terminal degradation. Our study now provides novel insights into how different VEGF-A isoforms can bind the same receptor tyrosine kinase and elicit diverse cellular outcomes.
Sigma-Aldrich
Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1054) Antibody, clone D1W, rabbit monoclonal, culture supernatant, clone D1W, Upstate®
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture