Merck
  • Discovery of a piperidine-4-carboxamide CCR5 antagonist (TAK-220) with highly potent Anti-HIV-1 activity.
Discovery of a piperidine-4-carboxamide CCR5 antagonist (TAK-220) with highly potent Anti-HIV-1 activity.
Journal of medicinal chemistry (2006-04-28)
Shinichi Imamura, Takashi Ichikawa, Youichi Nishikawa, Naoyuki Kanzaki, Katsunori Takashima, Shinichi Niwa, Yuji Iizawa, Masanori Baba, Yoshihiro Sugihara
ABSTRACT
We incorporated various polar groups into previously described piperidine-4-carboxamide CCR5 antagonists to improve their metabolic stability in human hepatic microsomes. Introducing a carbamoyl group into the phenyl ring of the 4-benzylpiperidine moiety afforded the less lipophilic compound 5f, which possessed both high metabolic stability and good inhibitory activity of HIV-1 envelope-mediated membrane fusion (IC(50) = 5.8 nM). Further optimization to increase potency led to the discovery of 1-acetyl-N-{3-[4-(4-carbamoylbenzyl)piperidin-1-yl]propyl}-N-(3-chloro-4-methylphenyl)piperidine-4-carboxamide (5m, TAK-220), which showed high CCR5 binding affinity (IC(50) = 3.5 nM) and potent inhibition of membrane fusion (IC(50) = 0.42 nM), as well as good metabolic stability. Compound 5m strongly inhibited the replication of CCR5-using HIV-1 clinical isolates in human peripheral blood mononuclear cells (mean EC(50) = 1.1 nM, EC(90) = 13 nM) and exhibited a good pharmacokinetic profile in monkeys (BA = 29%). This compound has been chosen as a clinical candidate for further development.
Sigma-Aldrich
4-(N-Boc-amino)piperidine, 96%
Pricing and availability is not currently available.