Merck
  • Elevated ratio of MMP2/MMP9 activity is associated with poor response to chemotherapy in osteosarcoma.
Elevated ratio of MMP2/MMP9 activity is associated with poor response to chemotherapy in osteosarcoma.
BMC cancer (2016-03-17)
Pierre Kunz, Heiner Sähr, Burkhard Lehner, Christian Fischer, Elisabeth Seebach, Jörg Fellenberg
ABSTRACT
Matrix metalloproteinases (MMPs) are crucially involved in the regulation of multiple stages of cancer progression. Elevated MMP levels have been associated with the development of metastases and poor prognosis in several types of cancer. However, the role of MMPs in osteosarcoma and their prognostic value is still unclear. Available data are conflicting, most likely due to different technical approaches. We hypothesized that in contrast to total mRNA or protein levels frequently analyzed in previous studies the enzymatic activities of MMPs and their inhibitors the tissue inhibitors of matrix metalloproteinases (TIMPs) are closer related to their biological functions. We therefore aimed to evaluate the reliability of different zymography techniques for the quantification of MMP and TIMP activities in osteosarcoma biopsies in order to investigate their distribution, possible regulation and prognostic value. All analyses were done using cryo-conserved osteosarcoma pretreatment biopsies (n = 18). Gene and protein expression of MMPs and TIMPs were analyzed by RT-qPCR and western blot analysis, respectively. Overall MMP activity was analyzed by in situ zymography, individual MMP activities were analyzed by gelatin zymography. Reverse zymography was used to detect and quantify TIMP activities. Strong overall MMP activities could be detected in osteosarcoma pretreatment biopsies with MMP2 and MMP9 as predominant active MMPs. In contrast to total RNA or protein expression MMP2 and MMP9 activities showed significant quantitative differences between good and poor responders. While MMP9 activity was high in the good responder group and significantly decreased in the poor responder group, MMP2 activity showed a reverse distribution. Likewise, significant differences were detected concerning the activity of TIMPs resulting in a negative correlation of TIMP1 activity with MMP2 activity (p = 0.044) and negative correlations of TIMP2 and TIMP3 with MMP9 activity (p = 0.007 and p = 0.006). In contrast to mRNA or protein levels MMP and TIMP activities showed significant differences between the analyzed good and poor responder groups. A shift from MMP9 to predominant MMP2 activity is associated with poor response to chemotherapy suggesting that the ratio of MMP2/MMP9 activity might be a valuable and easily accessible marker to predict the response to chemotherapy in osteosarcoma.
Sigma-Aldrich
TIMP1 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)