Merck
  • TRPV1 on astrocytes rescues nigral dopamine neurons in Parkinson's disease via CNTF.
TRPV1 on astrocytes rescues nigral dopamine neurons in Parkinson's disease via CNTF.
Brain : a journal of neurology (2015-10-23)
Jin H Nam, Eun S Park, So-Yoon Won, Yu A Lee, Kyoung I Kim, Jae Y Jeong, Jeong Y Baek, Eun J Cho, Minyoung Jin, Young C Chung, Byoung D Lee, Sung Hyun Kim, Eung-Gook Kim, Kyunghee Byun, Bonghee Lee, Dong Ho Woo, C Justin Lee, Sang R Kim, Eugene Bok, Yoon-Seong Kim, Tae-Beom Ahn, Hyuk Wan Ko, Saurav Brahmachari, Olga Pletinkova, Juan C Troconso, Valina L Dawson, Ted M Dawson, Byung K Jin
ABSTRACT
Currently there is no neuroprotective or neurorestorative therapy for Parkinson's disease. Here we report that transient receptor potential vanilloid 1 (TRPV1) on astrocytes mediates endogenous production of ciliary neurotrophic factor (CNTF), which prevents the active degeneration of dopamine neurons and leads to behavioural recovery through CNTF receptor alpha (CNTFRα) on nigral dopamine neurons in both the MPP(+)-lesioned or adeno-associated virus α-synuclein rat models of Parkinson's disease. Western blot and immunohistochemical analysis of human post-mortem substantia nigra from Parkinson's disease suggests that this endogenous neuroprotective system (TRPV1 and CNTF on astrocytes, and CNTFRα on dopamine neurons) might have relevance to human Parkinson's disease. Our results suggest that activation of astrocytic TRPV1 activates endogenous neuroprotective machinery in vivo and that it is a novel therapeutic target for the treatment of Parkinson's disease.
Sigma-Aldrich
Rinvanil, ≥98% (HPLC)
Pricing and availability is not currently available.