Merck
  • Moxifloxacin resistance in the F15/LAM4/KZN extensively drug-resistant strain of Mycobacterium tuberculosis.
Moxifloxacin resistance in the F15/LAM4/KZN extensively drug-resistant strain of Mycobacterium tuberculosis.
Infection and drug resistance (2014-09-05)
Navisha Dookie, A Willem Sturm, Prashini Moodley
ABSTRACT
Moxifloxacin (MXF) has been advocated for the treatment of extensively drug-resistant (XDR) tuberculosis despite resistance to older-generation fluoroquinolones. We investigated the relationship between the minimum inhibitory concentration (MIC) of MXF and mutations in the gyrA and gyrB genes in Mycobacterium tuberculosis (MTB) isolates from KwaZulu-Natal (KZN) Province of South Africa. MICs of 56 MTB isolates were compared to the mutations in the quinolone resistance-determining region known to confer fluoroquinolone resistance. Isolates were genotyped by IS6110 restriction fragment length polymorphism analysis. The circulating F15/LAM4/KZN XDR strain circulating in KZN Province harbored the A90V mutation and displayed high-level resistance with MICs of 8 mg/L for ciprofloxacin and ofloxacin and ≥1 mg/L for MXF. The inclusion of MXF in XDR-TB treatment regimens requires careful consideration in our setting, where clinical outcome data in MXF-containing regimens are unavailable.
Product Description
Roche
Expand High Fidelity PCR System, dNTPack, suitable for PCR, High Fidelity PCR, hotstart: no, dNTPs included
Pricing and availability is not currently available.