Merck
  • High prevalence and infection levels of Nosema ceranae in bumblebees Bombus atratus and Bombus bellicosus from Uruguay.
High prevalence and infection levels of Nosema ceranae in bumblebees Bombus atratus and Bombus bellicosus from Uruguay.
Journal of invertebrate pathology (2015-08-08)
N Arbulo, K Antúnez, S Salvarrey, E Santos, B Branchiccela, R Martín-Hernández, M Higes, C Invernizzi
ABSTRACT
Nosema ceranae is one of the most prevalent pathogens in Apis mellifera and has recently been found in multiple host species including several species of bumblebees. Prevalence and infection intensity of N. ceranae was determined in two species of native bumblebees from Uruguay. Nosema ceranae was the only microsporidia identified and mean prevalence was 72% in Bombus atratus and 63% in Bombus bellicosus, values much higher than those reported elsewhere. The presence of this pathogen in bumblebees may be threatening not only for bumblebee populations, but also to the rest of the native pollinator community and to honeybees.
Product Description
Roche
FastStart PCR Master, sufficient for ≤100 reactions (04710436001), sufficient for ≤400 reactions (04710444001), sufficient for ≤2,000 reactions (04710452001)
Pricing and availability is not currently available.