Merck
Cell motion predicts human epidermal stemness.
The Journal of cell biology (2015-04-22)
Daisuke Nanba, Fujio Toki, Sota Tate, Matome Imai, Natsuki Matsushita, Ken Shiraishi, Koji Sayama, Hiroshi Toki, Shigeki Higashiyama, Yann Barrandon
ABSTRACT
Image-based identification of cultured stem cells and noninvasive evaluation of their proliferative capacity advance cell therapy and stem cell research. Here we demonstrate that human keratinocyte stem cells can be identified in situ by analyzing cell motion during their cultivation. Modeling experiments suggested that the clonal type of cultured human clonogenic keratinocytes can be efficiently determined by analysis of early cell movement. Image analysis experiments demonstrated that keratinocyte stem cells indeed display a unique rotational movement that can be identified as early as the two-cell stage colony. We also demonstrate that α6 integrin is required for both rotational and collective cell motion. Our experiments provide, for the first time, strong evidence that cell motion and epidermal stemness are linked. We conclude that early identification of human keratinocyte stem cells by image analysis of cell movement is a valid parameter for quality control of cultured keratinocytes for transplantation.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-8, clone ASC-8, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone 3E1, ascites fluid, clone 3E1, Chemicon®