Merck
  • Methylenetetrahydrofolate reductase gene polymorphisms in children with attention deficit hyperactivity disorder.
Methylenetetrahydrofolate reductase gene polymorphisms in children with attention deficit hyperactivity disorder.
International journal of medical sciences (2011-09-08)
Cem Gokcen, Nadir Kocak, Ahmet Pekgor
ABSTRACT
The purpose of this study was to evaluate the relationship between 5,10- methylenetetrahydrofolate reductase (MTHFR) polymorphisms and Attention Deficit Hyperactivity Disorder (ADHD) in a sample of Turkish children. MTHFR gene polymorphisms were assessed in 40 patients with ADHD and 30 healty controls. Two mutations in the MTHFR gene were investigated using polymerase chain reactions and restriction fragment length polymorphisms. Although there were no statistically significant differences in genotype distributions of the C677T alleles between the ADHD and the control groups (p=0,678) but the genotypic pattern of the distributions of the A1298C alleles was different between the ADHD patients and the controls (p=0,033). Preliminary data imply a possible relationship between A1298C MTHFR polymorphisms and the ADHD.
Sigma-Aldrich
10X PCR Buffer, Optimized for routine PCR with MgCl2 included
Pricing and availability is not currently available.