Merck
A concise total synthesis of DL-histrionicotoxin.
Journal of the American Chemical Society (2006-09-28)
Maheswaran S Karatholuvhu, Alex Sinclair, Annabella F Newton, Marie-Lyne Alcaraz, Robert A Stockman, Philip L Fuchs
ABSTRACT
The synthesis of (+/-)-histrionicotoxin has been achieved in just nine steps using a two-directional synthesis strategy. Key reactions include a two-directional cross-metathesis, a tandem oxime formation/Michael addition/1,4-prototopic shift/[3 + 2]-cycloaddition cascade, a selective Z,Z-bisenyne formation, and a one-pot N-O and bischloroacetylene reduction.
Sigma-Aldrich
5-Bromo-1-pentene, 95%