Merck
  • Catalytic asymmetric synthesis with rh-diene complexes: 1,4-addition of arylboronic acids to unsaturated esters.
Catalytic asymmetric synthesis with rh-diene complexes: 1,4-addition of arylboronic acids to unsaturated esters.
Organic letters (2005-08-12)
Jean-François Paquin, Corey R J Stephenson, Christian Defieber, Erick M Carreira
ABSTRACT
A general route to enantioenriched tert-butyl 3,3-diarylpropanoates is presented. These useful building blocks are prepared via an asymmetric rhodium-catalyzed conjugate addition of arylboronic acids to unsaturated tert-butyl esters in the presence of chiral dienes as ligands. The addition of both electron-poor and electron-rich boronic acids proceeds smoothly with various enoates in 63-90% yield with high enantioselectivites (89-94% ee). [reaction: see text]