Merck
  • 2-Dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole: a novel powerful and selective inhibitor of protein kinase CK2.
2-Dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole: a novel powerful and selective inhibitor of protein kinase CK2.
Biochemical and biophysical research communications (2004-09-11)
Mario A Pagano, Flavio Meggio, Maria Ruzzene, Mariola Andrzejewska, Zygmunt Kazimierczuk, Lorenzo A Pinna
ABSTRACT
Protein kinase CK2 is a highly pleiotropic enzyme whose high constitutive activity is suspected to be instrumental to the enhancement of the tumour phenotype and to the propagation of infectious diseases. Here we describe a novel compound, 2-dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole (DMAT), which is superior to the commonly used specific CK2 inhibitor 4,5,6,7-tetrabromobenzotriazole (TBB) in several respects. DMAT displays the lowest K(i) value ever reported for a CK2 inhibitor (40 nM); it is cell permeable and its efficacy on cultured cells, both in terms of endogenous CK2 inhibition and induction of apoptosis, is several fold higher than that of TBB. The selectivity of DMAT assayed on a panel of >30 protein kinases is comparable to that of TBB, with the additional advantage of being ineffective on protein kinase CK1 up to 200 microM. These properties make DMAT the first choice CK2 inhibitor for in vivo studies available to date.
Sigma-Aldrich
DMAT, ≥98% (HPLC)
Pricing and availability is not currently available.