Merck
Nanoparticle amplification
Journal of materials chemistry. B (2013-11-02)
Justin H Lo, Geoffrey von Maltzahn, Jacqueline Douglass, Ji-Ho Park, Michael J Sailor, Erkki Ruoslahti, Sangeeta N Bhatia
ABSTRACT
The success of nanoparticle-based cancer therapies ultimately depends on their ability to selectively and efficiently accumulate in regions of disease. Outfitting nanoparticles to actively target tumor-specific markers has improved specificity, yet it remains a challenge to amass adequate therapy in a selective manner. To help address this challenge, we have developed a mechanism of nanoparticle amplification based on stigmergic (environment-modifying) signalling, in which a "Signalling" population of gold nanorods induces localized unveiling of cryptic collagen epitopes, which are in turn targeted by "Responding" nanoparticles bearing gelatin-binding fibronectin fragments. We demonstrate that this two-particle system results in significantly increased, selective recruitment of responding particles. Such amplification strategies have the potential to overcome limitations associated with single-particle targeting by leveraging the capacity of nanoparticles to interact with their environment to create abundant new binding motifs.
Sigma-Aldrich
Fibronectin Proteolytic Fragment from human plasma, lyophilized powder, 45 kDa