Merck
Synthesis of (R)- and (S)- muscone.
Bioscience, biotechnology, and biochemistry (2002-08-07)
Satoko Fujimoto, Keisuke Yoshikawa, Masamichi Itoh, Takeshi Kitaharai
ABSTRACT
(R)-(-)-Muscone (3-methylcyclopentadecanone, 1) the key perfumery component isolated from the male musk deer, Moschus moschiferus,* was synthesized from the easily available chiral building block, (R)-3-tert-butoxycarbonyl-2-methylpropanoic acid (2), by employing ring-closing olefin metathesis (RCM). Antipode (+)-1 was also synthesized in a similar manner from tert-butyl (S)-3-methoxycarbonylbutanoate (10). *(a) Walbaum, H. J. J. Prakt. Chem., 73, 488 (1906); (b) Ruzicka, L., Further considerations on the constitution of muscone. Helv. Chim. Acta, 9, 715, 1008-1017 (1926).
Sigma-Aldrich
Indium(III) chloride, 98%
Pricing and availability is not currently available.