Merck
  • Beta-proline analogues as agonists at the strychnine-sensitive glycine receptor.
Beta-proline analogues as agonists at the strychnine-sensitive glycine receptor.
Journal of medicinal chemistry (1992-01-24)
G Johnson, J T Drummond, P A Boxer, R F Bruns
ABSTRACT
3-Carboxy-3,4-dehydropyrrolidine was found to bind with affinity equal to that of glycine in a [3H]strychnine binding assay. Simple substitution of the 1-, 2-, 4-, or 5-position resulted in marked loss of affinity. A decline in affinity was also found upon enlargement, contraction, or saturation of the 5-membered ring. However, beta-proline and azetidine-3-carboxylic acid retained significant binding affinity. Despite its good affinity in [3H]strychnine binding, 3-carboxy-3,4-dehydropyrrolidine showed only weak agonist activity in intracellular recordings of cultured murine spinal cord neurons. This apparent lack of correlation between binding and functional results is discussed in light of the current models of the strychnine-sensitive glycine receptor.
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
3-Azetidinecarboxylic acid, 98%
Pricing and availability is not currently available.