Merck
  • Enzymatic synthesis of [4R-2H]NAD (P)H and [4S-2H]NAD(P)H and determination of the stereospecificity of 7 alpha- and 12 alpha hydroxysteroid dehydrogenase.
Enzymatic synthesis of [4R-2H]NAD (P)H and [4S-2H]NAD(P)H and determination of the stereospecificity of 7 alpha- and 12 alpha hydroxysteroid dehydrogenase.
Biochimica et biophysica acta (1989-10-05)
G Ottolina, S Riva, G Carrea, B Danieli, A F Buckmann
ABSTRACT
The stereospecifically labeled coenzymes [4R-2H]NADH, [4R-2H]NADPH and [4S-2H]NAD(P)H were synthesized enzymatically in high yield and high isotopic purity (greater than or equal to 95%) with 2HCOO2H/formate dehydrogenase, (CH3)2C2HOH/alchol dehydrogenase from Thermoanaerobium brockii and [1-2H]glucose/glucose dehydrogenase, respectively. This set of deuterated coenzymes was used to determine the stereospecificity of the previously unstudied 7 alpha-hydroxysteroid dehydrogenase from Escherichia coli (NAD-dependent) and 12 alpha-hydroxysteroid dehydrogenase from Clostridium group P (NADP-dependent). H-NMR and EI-MS of the nicotinamide moiety after enzymatic oxidation of deuterated NAD(P)H with dehydrocholic acid as substrate showed that both dehydrogenases are B-sterospecific.
Sigma-Aldrich
Formic acid-d2, 95 wt. % in D2O, 98 atom % D
Pricing and availability is not currently available.