Merck
  • Synthesis of novel Se-substituted selenocysteine derivatives as potential kidney selective prodrugs of biologically active selenol compounds: evaluation of kinetics of beta-elimination reactions in rat renal cytosol.
Synthesis of novel Se-substituted selenocysteine derivatives as potential kidney selective prodrugs of biologically active selenol compounds: evaluation of kinetics of beta-elimination reactions in rat renal cytosol.
Journal of medicinal chemistry (1996-05-10)
I Andreadou, W M Menge, J N Commandeur, E A Worthington, N P Vermeulen
ABSTRACT
Eighteen Se-substituted selenocysteine derivatives were synthesized as potential kidney selective prodrugs which can be activated by renal cysteine conjugate beta-lyase to selenium-containing chemoprotectants or antitumor agents. Selenocysteine derivatives with aliphatic and benzylic Se-substituents were synthesized by reducing selenocystine to selenocysteine followed by a reaction with the corresponding alkyl and benzyl halogenides. Selenocysteine derivatives with aromatic Se-substitutes were synthesized by reaction of beta-chloroalanine with substituted phenylselenol compounds, which were formed by reducing substituted diphenyl diselenides by NaBH4. The enzyme kinetic parameters (apparent Km and Vmax) of the beta-elimination reaction of the selenocysteine conjugates were studied in rat renal cytosol. The results suggest that Se-substituted L-selenocysteine conjugates are extremely good substrates for renal cysteine conjugate beta-lyases as indicated by low apparent Km and high Vmax values. The benzyl-substituted Se-conjugates appeared to be better substrates than the phenyl- and alkyl-substituted Se-conjugates. Corresponding L-cysteine S-conjugates were too poor substrates to obtain proper enzyme kinetics. Recently, local activation of cysteine S-conjugates by renal cysteine conjugate beta-lyases was proposed as a new strategy to target antitumor agents to the kidney. The present results show that Se-substituted selenocysteine conjugates may be more promising prodrugs because these compounds are much better substrates for beta-lyase.
Sigma-Aldrich
Seleno-L-cystine, 95%
Pricing and availability is not currently available.