Merck
  • MicroRNAs cooperatively inhibit a network of tumor suppressor genes to promote pancreatic tumor growth and progression.
MicroRNAs cooperatively inhibit a network of tumor suppressor genes to promote pancreatic tumor growth and progression.
Gastroenterology (2013-10-15)
Adam E Frampton, Leandro Castellano, Teresa Colombo, Elisa Giovannetti, Jonathan Krell, Jimmy Jacob, Loredana Pellegrino, Laura Roca-Alonso, Niccola Funel, Tamara M H Gall, Alexander De Giorgio, Filipa G Pinho, Valerio Fulci, David J Britton, Raida Ahmad, Nagy A Habib, R Charles Coombes, Victoria Harding, Thomas Knösel, Justin Stebbing, Long R Jiao
ABSTRACT
There has not been a broad analysis of the combined effects of altered activities of microRNAs (miRNAs) in pancreatic ductal adenocarcinoma (PDAC) cells, and it is unclear how these might affect tumor progression or patient outcomes. We combined data from miRNA and messenger RNA (mRNA) expression profiles and bioinformatic analyses to identify an miRNA-mRNA regulatory network in PDAC cell lines (PANC-1 and MIA PaCa-2) and in PDAC samples from patients. We used this information to identify miRNAs that contribute most to tumorigenesis. We identified 3 miRNAs (MIR21, MIR23A, and MIR27A) that acted as cooperative repressors of a network of tumor suppressor genes that included PDCD4, BTG2, and NEDD4L. Inhibition of MIR21, MIR23A, and MIR27A had synergistic effects in reducing proliferation of PDAC cells in culture and growth of xenograft tumors in mice. The level of inhibition was greater than that of inhibition of MIR21 alone. In 91 PDAC samples from patients, high levels of a combination of MIR21, MIR23A, and MIR27A were associated with shorter survival times after surgical resection. In an integrated data analysis, we identified functional miRNA-mRNA interactions that contribute to growth of PDACs. These findings indicate that miRNAs act together to promote tumor progression; therapeutic strategies might require inhibition of several miRNAs.
Sigma-Aldrich
Anti-NEDD4L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-SORBS2 antibody,Mouse monoclonal, clone S5C, purified from hybridoma cell culture
Pricing and availability is not currently available.