Merck
  • Interleukin 18--binding protein ameliorates liver ischemia--reperfusion injury.
Interleukin 18--binding protein ameliorates liver ischemia--reperfusion injury.
The Journal of surgical research (2016-02-07)
Ahmet Bal, Yucel Gonul, Omer Hazman, Ahmet Kocak, Mehmet Fatih Bozkurt, Sezgin Yilmaz, Serdar Kokulu, Oya Oruc, Kasim Demir
ABSTRACT
The aim of this study was to investigate the possible protective effect of interleukin 18-binding protein (IL-18BP) on ischemia-reperfusion (I/R)-induced liver injury in experimental rat models. Liver is one of the most affected organs from I/R process. IL-18 is an important proinflammatory cytokine, which may induce some events such as production of reactive oxygen substances and release of various cytokines. IL-18BP acts as an inhibitor of IL-18. The relationship between IL-18 and IL-18BP has an important place in inflammatory process. Rats were equally divided into three groups as follows: sham: Hepatic pedicle dissection was done, but hepatic pedicle clamping was not used. I/R: Sixty minutes of ischemia and 2 h of reperfusion were applied. IR + IL-18BP: Recombinant human IL-18BP (100 μg/kg) was administered 30 min before the surgery. Hepatic pedicle was clamped during 60 min of ischemia and 2 h of reperfusion was achieved. Liver enzyme levels were significantly lower in the IR + IL-18BP group, when compared with the I/R group. Serum and tissue levels of tumor necrosis factor-α, IL-6, and IL-18 were considerably lower in the IR + IL-18BP group, when compared with the I/R group, but hepatic interferon-γ and IL1β levels were not significant. Serum oxidative stress index level was significantly higher in the I/R group, when compared with the IR + IL-18BP group. In immunostaining, it was observed that pathologic changes were lower in IR + IL-18BP group than the I/R group. IL-18BP exhibited anti-inflammatory, antioxidant, and protective effects in I/R-mediated hepatic injury via regulating some liver enzyme activities and cytokine levels. Additionally, these effects have been verified by histomorphologic examination and oxidative stress markers.
Sigma-Aldrich
IL18BP human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)