Merck
  • Novel mannich bases, 5-arylimidazolidine-2,4-dione derivatives with dual 5-HT(1A) receptor and serotonin transporter affinity.
Novel mannich bases, 5-arylimidazolidine-2,4-dione derivatives with dual 5-HT(1A) receptor and serotonin transporter affinity.
Archiv der Pharmazie (2013-01-05)
Anna Czopek, Marcin Kołaczkowski, Adam Bucki, Hanna Byrtus, Maciej Pawłowski, Agata Siwek, Andrzej J Bojarski, Marek Bednarski, Dagmara Wróbel, Anna Wesołowska
ABSTRACT
A computer aided ligand design study of imidazolidine-2,4-dione derivatives was conducted in order to obtain compounds with dual 5-HT(1A) receptor and serotonin transporter (SERT) affinity. According to molecular modeling results, series of Mannich bases were chosen and synthesized. Investigated compounds were tested for 5-HT(1A) , 5-HT(2A) , α(1) and SERT affinity. Two selected compounds (5, 9) were characterized in functional experiments and possessed a pharmacological profile which may enhance SERT blocking efficacy - 5-HT(1A) partial agonism and 5-HT(2A) antagonism in one molecule. Furthermore these compounds displayed satisfactory selectivity over adrenergic α(1) receptors. The most promising compounds, 5-arylimidazolidine-2,4-dione derivatives with 4-(3-chlorophenyl)piperazinylmethyl moiety were tested for antidepressant and anxiolytic activity. In particular, compound 5 (5-(2-methoxyphenyl)-3-{1-[4-(3-chlorophenyl)piperazin-1-yl]methyl}-imidazolidine-2,4-dione), tested in the forced swim test in mice, exhibited a favorable antidepressant-like profile without affecting spontaneous locomotor activity.