Merck
  • Synthesis and structure-activity relationships of 2-phenyl-1-[(pyridinyl- and piperidinylmethyl)amino]-3-(1H-1,2,4-triazol-1-yl)propan-2-ols as antifungal agents.
Synthesis and structure-activity relationships of 2-phenyl-1-[(pyridinyl- and piperidinylmethyl)amino]-3-(1H-1,2,4-triazol-1-yl)propan-2-ols as antifungal agents.
Bioorganic & medicinal chemistry letters (2008-12-19)
Francis Giraud, Rémi Guillon, Cédric Logé, Fabrice Pagniez, Carine Picot, Marc Le Borgne, Patrice Le Pape
ABSTRACT
Continuous efforts on the synthesis and structure-activity relationships (SARs) studies of modified 1-benzylamino-2-phenyl-3-(1H-1,2,4-triazol-1-yl)propan-2-ols as antifungal agents, allowed identification of new 1-[(pyridinyl- and piperidinylmethyl)amino] derivatives with MIC(80) values ranging from 1410.0 to 23.0ngmL(-1) on Candidaalbicans. These results confirmed both the importance of pi-pi stacking and hydrogen bonding interactions in the active site of CYP51-C. albicans.
Sigma-Aldrich
4-Piperidinemethanol, 97%
Pricing and availability is not currently available.