Merck
  • Combined effects of pollutants and salinity on embryo-larval development of the Pacific oyster, Crassostrea gigas.
Combined effects of pollutants and salinity on embryo-larval development of the Pacific oyster, Crassostrea gigas.
Marine environmental research (2015-11-20)
Perrine Gamain, Patrice Gonzalez, Jérôme Cachot, Patrick Pardon, Nathalie Tapie, Pierre Yves Gourves, Hélène Budzinski, Bénédicte Morin
ABSTRACT
For several years, low larval recruitment has been observed in Arcachon Bay, in southwest France. Exposure to pollutants could partly account for the reduction of early life stages of the Pacific oyster. This study evaluated the effects of copper and S-metolachlor in combination with salinity on the early life stages of Crassostrea gigas. Embryos were exposed to concentrations of copper (1, 10 and 50 μg L(-1)) or S-metolachlor (10, 100 and 1000 ng L(-1)) and six salinities (18, 21, 24, 27, 30 and 33 u.s.i). Embryotoxicity was measured by considering both the percentage of abnormalities and arrested development in D-shaped larvae. Embryo-larval development was only affected at salinities ≤24 u.s.i, which have never been observed during C. gigas reproduction period in Arcachon Bay. Both contaminants had an effect at environmental concentrations. Our results suggest that copper and metolachlor toxicity was enhanced with decreasing salinity.
Product Description
Supelco
Metolachlor, predominantly S-Metolachlor, PESTANAL®, analytical standard
Pricing and availability is not currently available.