Merck
  • Thermally Driven Large N-Type Voltage Responses from Hybrids of Carbon Nanotubes and Poly(3,4-ethylenedioxythiophene) with Tetrakis(dimethylamino)ethylene.
Thermally Driven Large N-Type Voltage Responses from Hybrids of Carbon Nanotubes and Poly(3,4-ethylenedioxythiophene) with Tetrakis(dimethylamino)ethylene.
Advanced materials (Deerfield Beach, Fla.) (2015-10-02)
Hong Wang, Jui-Hung Hsu, Su-In Yi, Suk Lae Kim, Kyungwho Choi, Gang Yang, Choongho Yu
ABSTRACT
Hybrids of carbon nanotubes (CNTs) and poly(3,4-ethylenedioxythiophene) (PEDOT) treated by tetrakis(dimethylamino)ethylene (TDAE) have large n-type voltages in response to temperature differences. The reduced carrier concentration by TDAE reduction and partially percolated CNT networks embedded in the PEDOT matrix result in high thermopower and low thermal conductivity. The high electron mobility in the CNTs helps to minimally reduce the electrical conductivity of the hybrid, resulting in a large figure-of-merit.
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrakis(dimethylamino)ethylene
Pricing and availability is not currently available.