Merck
  • Combinatorial peptidomics: a generic approach for protein expression profiling.
Combinatorial peptidomics: a generic approach for protein expression profiling.
Journal of nanobiotechnology (2003-07-25)
Mikhail Soloviev, Richard Barry, Elaine Scrivener, Jonathan Terrett
ABSTRACT
Traditional approaches to protein profiling were built around the concept of investigating one protein at a time and have long since reached their limits of throughput. Here we present a completely new approach for comprehensive compositional analysis of complex protein mixtures, capable of overcoming the deficiencies of current proteomics techniques. The Combinatorial methodology utilises the peptidomics approach, in which protein samples are proteolytically digested using one or a combination of proteases prior to any assay being carried out. The second fundamental principle is the combinatorial depletion of the crude protein digest (i.e. of the peptide pool) by chemical crosslinking through amino acid side chains. Our approach relies on the chemical reactivities of the amino acids and therefore the amino acid content of the peptides (i.e. their information content) rather than their physical properties. Combinatorial peptidomics does not use affinity reagents and relies on neither chromatography nor electrophoretic separation techniques. It is the first generic methodology applicable to protein expression profiling, that is independent of the physical properties of proteins and does not require any prior knowledge of the proteins. Alternatively, a specific combinatorial strategy may be designed to analyse a particular known protein on the basis of that protein sequence alone or, in the absence of reliable protein sequence, even the predicted amino acid translation of an EST sequence. Combinatorial peptidomics is especially suitable for use with high throughput micro- and nano-fluidic platforms capable of running multiple depletion reactions in a single disposable chip.