Merck
  • Purification and structural determination of SCB1, a gamma-butyrolactone that elicits antibiotic production in Streptomyces coelicolor A3(2).
Purification and structural determination of SCB1, a gamma-butyrolactone that elicits antibiotic production in Streptomyces coelicolor A3(2).
The Journal of biological chemistry (2001-02-07)
E Takano, T Nihira, Y Hara, J J Jones, C J Gershater, Y Yamada, M Bibb
ABSTRACT
Early stationary phase culture supernatants of Streptomyces coelicolor A3(2) contained at least four small diffusible signaling molecules that could elicit precocious antibiotic synthesis in the producing strain. The compounds were not detected in exponentially growing cultures. One of these compounds, SCB1, was purified to homogeneity and shown to be a gamma-butyrolactone of structure (2R, 3R,1'R)-2-(1'-hydroxy-6-methylheptyl)-3-hydroxymethylbutanolide . Bioassays of chemically synthesized SCB1, and of its purified stereoisomers, suggest that SCB1 acts in a highly specific manner to elicit the production of both actinorhodin and undecylprodigiosin, the two pigmented antibiotics made by S. coelicolor.
Sigma-Aldrich
1-Bromo-4-methylpentane, 97%
Pricing and availability is not currently available.