Merck
Sequential photo-oxidation of methanol to methyl formate on TiO2(110).
Journal of the American Chemical Society (2012-12-28)
Katherine R Phillips, Stephen C Jensen, Martin Baron, Shao-Chun Li, Cynthia M Friend
ABSTRACT
Methyl formate is produced from the photo-oxidation of methanol on preoxidized TiO(2)(110). We demonstrate that two consecutive photo-oxidation steps lead to methyl formate using mass spectrometry and scanning tunneling microscopy. The first step in methanol oxidation is formation of methoxy by the thermal dissociation of the O-H bond to yield adsorbed CH(3)O and water. Formaldehyde is produced via hole-mediated oxidation of adsorbed methoxy in the first photochemical step. Next, transient HCO is made photochemically from formaldehyde. The HCO couples with residual methoxy on the surface to yield methyl formate. Exposure of the titania surface to O(2) is required for these photo-oxidation steps in order to heal surface and near-surface defects that can serve as hole traps. Notably, residual O adatoms are not required for photochemical production of methyl formate or formaldehyde. All O adatoms react thermally with methanol to form methoxy and gaseous water at rt, leaving a surface devoid of O adatoms. The mechanism provides insight into the photochemistry of TiO(2) and suggests general synthetic pathways that are the result of the ability to activate both alkoxides and aldehydes using photons.
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl formate, spectrophotometric grade, ≥98%
Pricing and availability is not currently available.