Merck
  • Improving expression of recombinant human IGF-1 using IGF-1R knockout CHO cell lines.
Improving expression of recombinant human IGF-1 using IGF-1R knockout CHO cell lines.
Biotechnology and bioengineering (2015-11-03)
Sandrine Romand, Thomas Jostock, Mara Fornaro, Joerg Schmidt, Anett Ritter, Burkhard Wilms, Holger Laux
ABSTRACT
Chinese Hamster Ovary (CHO) cells are widely used for the large-scale production of recombinant biopharmaceuticals. However, attempts to express IGF-1 (a mutated human Insulin-like growth factor 1 Ea peptide (hIGF-1Ea mut)) in CHO cells resulted in poor cell growth and low productivity (0.1-0.2 g/L). Human IGF-1 variants negatively impacted CHO cell growth via the IGF-1 receptor (IGF-1R). Therefore knockout (KO) of the IGF-1R gene in two different CHO cell lines as well as knockdown (KD) of IGF-1R in one CHO cell line were performed. These cell line engineering approaches decreased significantly the hIGF-1 mediated cell growth inhibition and increased productivity of both KO CHO cell lines as well as of the KD CHO cell line. A productivity increase of 10-fold at pool level and sevenfold at clone level was achieved, resulting in a titer of 1.3 g/L. This data illustrate that cell line engineering approaches are powerful tools to improve the yields of recombinant proteins which are difficult to produce in CHO cells.
Sigma-Aldrich
Extract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
Extract-N-Amp Blood PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Pricing and availability is not currently available.
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Pricing and availability is not currently available.