Merck
  • PDK1 signaling toward PLK1-MYC activation confers oncogenic transformation, tumor-initiating cell activation, and resistance to mTOR-targeted therapy.
PDK1 signaling toward PLK1-MYC activation confers oncogenic transformation, tumor-initiating cell activation, and resistance to mTOR-targeted therapy.
Cancer discovery (2013-07-28)
Jing Tan, Zhimei Li, Puay Leng Lee, Peiyong Guan, Mei Yee Aau, Shuet Theng Lee, Min Feng, Cheryl Zihui Lim, Eric Yong Jing Lee, Zhen Ning Wee, Yaw Chyn Lim, R K Murthy Karuturi, Qiang Yu
ABSTRACT
Although 3-phosphoinositide-dependent protein kinase-1 (PDK1) has been predominately linked to the phosphoinositide 3-kinase (PI3K)-AKT pathway, it may also evoke additional signaling outputs to promote tumorigenesis. Here, we report that PDK1 directly induces phosphorylation of Polo-like kinase 1 (PLK1), which in turn induces MYC phosphorylation and protein accumulation. We show that PDK1-PLK1-MYC signaling is critical for cancer cell growth and survival, and small-molecule inhibition of PDK1/PLK1 provides an effective approach for therapeutic targeting of MYC dependency. Intriguingly, PDK1-PLK1-MYC signaling induces an embryonic stem cell-like gene signature associated with aggressive tumor behaviors and is a robust signaling axis driving cancer stem cell (CSC) self-renewal. Finally, we show that a PLK1 inhibitor synergizes with an mTOR inhibitor to induce synergistic antitumor effects in colorectal cancer by antagonizing compensatory MYC induction. These findings identify a novel pathway in human cancer and CSC activation and provide a therapeutic strategy for targeting MYC-associated tumorigenesis and therapeutic resistance. This work identifies PDK1–PLK1-MYC signaling as a new oncogenic pathway driving oncogenic transformation and CSC self-renewal. Targeted inhibition of PDK1/PLK1 is robust in targeting MYC dependency in cancer cells. Thus, our findings provide important insights into cancer and CSC biology and have significant therapeutic implications.
Sigma-Aldrich
PDK1, active, His tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.