Merck
  • Facile preparation of beta-fluoro amines by the reaction of aziridines with potassium fluoride dihydrate in the presence of Bu4NHSO4.
Facile preparation of beta-fluoro amines by the reaction of aziridines with potassium fluoride dihydrate in the presence of Bu4NHSO4.
The Journal of organic chemistry (2004-01-17)
Ren-Hua Fan, Yong-Gui Zhou, Wan-Xuan Zhang, Xue-Long Hou, Li-Xin Dai
ABSTRACT
Potassium fluoride combined with tetrabutylammonium bisulfate is an efficient reagent to convert a variety of aziridines derived from cyclic and acyclic alkenes to beta-fluoro amine derivatives in high yield.
Sigma-Aldrich
Potassium fluoride dihydrate, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O