Merck
  • Clinical determinants of reduced physical activity in hemodialysis and peritoneal dialysis patients.
Clinical determinants of reduced physical activity in hemodialysis and peritoneal dialysis patients.
Journal of nephrology (2014-12-17)
Gabriela Cobo, Paloma Gallar, Thiane Gama-Axelsson, Cristina Di Gioia, Abdul Rashid Qureshi, Rosa Camacho, Ana Vigil, Olof Heimbürger, Olimpia Ortega, Isabel Rodriguez, Juan Carlos Herrero, Peter Bárány, Bengt Lindholm, Peter Stenvinkel, Juan Jesús Carrero
ABSTRACT
The phenotype associated to reduced physical activity (PA) in dialysis patients is poorly documented. We here evaluate weekly PA in two independent cohorts. Cross-sectional study with PA assessed by the number of steps/day measured by pedometer in two cohorts of prevalent dialysis patients: (1) peritoneal dialysis (PD) patients (n = 64; 62 ± 14 years; 70 % men) from Stockholm, Sweden using the pedometer for 7 consecutive days; (2) hemodialysis (HD) patients (n = 78; 63 ± 12 years; 65% men) from a single center in Madrid, Spain using the pedometer for 6 consecutive days: 2 HD days, 2 non-HD midweek days and 2 non-HD weekend days. In both cohorts, comorbidities, body composition, nutritional status, and related biomarkers were assessed. Cohorts were not merged; instead data were analyzed separately serving as reciprocal replication analyses. Most patients (63% of PD and 71% of HD) were considered sedentary (<5,000 steps/day). PD patients had on average 4,839 ± 3,313 steps/day. HD patients had 3,767 ± 3,370 steps/day on HD-free days, but fewer steps/day on HD days (2,274 ± 2,048 steps/day; p < 0.0001). In both cohorts, and across increasing PA tertiles, patients were younger and had less comorbidities. Higher PA was also accompanied by better nutritional status (depicted by albumin, pre-albumin, creatinine and normalized protein catabolic rate in HD, and by albumin and subjective global assessment [SGA] in PD), higher lean body mass, and lower fat body mass (bioimpedance and/or dual-energy X-ray absorptiometry [DEXA]). Higher levels of PA were accompanied by lower levels of C-reactive protein in PD. Age and lean body mass were the strongest multivariate predictors of PA in both cohorts. There is a high prevalence of sedentary behavior in dialysis patients. Better physical activity was consistently associated with younger age, lower presence of comorbidities and better nutritional status. Pedometers represent a simple and inexpensive tool to objectively evaluate physical activity in this patient population.
Sigma-Aldrich
Carbendazim, 97%