Merck
  • Formononetin sensitizes glioma cells to doxorubicin through preventing EMT via inhibition of histone deacetylase 5.
Formononetin sensitizes glioma cells to doxorubicin through preventing EMT via inhibition of histone deacetylase 5.
International journal of clinical and experimental pathology (2015-08-12)
Quan Liu, Yan Sun, Jie-Min Zheng, Xian-Lei Yan, Hong-Mou Chen, Jia-Kang Chen, He-Qing Huang
ABSTRACT
Chemoresistance is a major obstacle to successful chemotherapy for glioma. Formononetin is a novel herbal isoflavonoid isolated from Astragalus membranaceus and possesses antitumorigenic properties. In the present study, we investigated the anti-proliferative effects of formononetin on human glioma cells, and further elucidated the molecular mechanism underlying the anti-tumor property. We found that formononetin enhanced doxorubicin cytotoxicity in glioma cells. Combined treatment with formononetin reversed the doxorubicin-induced epithelial-mesenchymal transition (EMT) in tumor cells. Moreover, we found that formononetin treatment significantly decreased the expression of HDAC5. Overexpression of HDAC5 diminished the suppressive effects of formononetin on glioma cell viability. Furthermore, knockdown of HDAC5 by siRNA inhibited the doxorubicin-induced EMT in glioma cells. Taken together, these results demonstrated that formononetin-combined therapy may enhance the therapeutic efficacy of doxorubicin in glioma cells by preventing EMT through inhibition of HDAC5.
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC5
Pricing and availability is not currently available.
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hdac5
Pricing and availability is not currently available.