Merck
  • Exogenous nitric oxide inhibits Rho-associated kinase activity in patients with angina pectoris: a randomized controlled trial.
Exogenous nitric oxide inhibits Rho-associated kinase activity in patients with angina pectoris: a randomized controlled trial.
Hypertension research : official journal of the Japanese Society of Hypertension (2015-03-06)
Tatsuya Maruhashi, Kensuke Noma, Noritaka Fujimura, Masato Kajikawa, Takeshi Matsumoto, Takayuki Hidaka, Ayumu Nakashima, Yasuki Kihara, James K Liao, Yukihito Higashi
ABSTRACT
The RhoA/Rho-associated kinase (ROCK) pathway has a key physiological role in the pathogenesis of atherosclerosis. Increased ROCK activity is associated with cardiovascular diseases. Endogenous nitric oxide (NO) has an anti-atherosclerotic effect, whereas the exogenous NO-mediated cardiovascular effect still remains controversial. The purpose of this study was to evaluate the effect of exogenous NO on ROCK activity in patients with angina pectoris. This is a prospective, open-label, randomized, controlled study. A total of 30 patients with angina pectoris were randomly assigned to receive 40 mg day(-1) of isosorbide mononitrate (n=15, 12 men and 3 women, mean age of 63±12 years, isosorbide mononitrate group) or conventional treatment (n=15, 13 men and 2 women, mean age of 64±13 years, control group) for 12 weeks. ROCK activity in peripheral leukocytes was measured by western blot analysis. ROCK activities at 4 and 12 weeks after treatment were decreased in the isosorbide mononitrate group (0.82±0.33 at 0 week, 0.62±0.20 at 4 weeks, 0.61±0.19 at 12 weeks, n=15 in each group, P<0.05, respectively) but not altered in the control group. ROCK1 and ROCK2 expression levels were similar in all treatment periods in the two groups. These findings suggest that the administration of exogenous NO can inhibit ROCK activity, indicating that the usage of exogenous NO could have a protective effect in patients with angina pectoris.
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline