Merck
High-alkali low-temperature polysulfide pulping (HALT) of Scots pine.
Bioresource technology (2015-06-30)
Markus Paananen, Herbert Sixta
ABSTRACT
High-alkali low-temperature polysulfide pulping (HALT) was effectively utilised to prevent major polysaccharide losses while maintaining the delignification rate. A yield increase of 6.7 wt% on wood was observed for a HALT pulp compared to a conventionally produced kappa number 60 pulp with comparable viscosity. Approximately 70% of the yield increase was attributed to improved galactoglucomannan preservation and 30% to cellulose. A two-stage oxygen delignification sequence with inter-stage peroxymonosulphuric acid treatment was used to ensure delignification to a bleachable grade. In a comparison to conventional pulp, HALT pulp effectively maintained its yield advantage. Diafiltration trials indicate that purified black liquor can be directly recycled, as large lignin fractions and basically all dissolved polysaccharides were separated from the alkali-rich BL.
Sigma-Aldrich
Potassium bisulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium bisulfate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Potassium bisulfate, JIS special grade, ≥98.0%