Merck
  • Multiplication of alpha-fetoprotein and protein induced by vitamin K absence-II is a powerful predictor of prognosis and recurrence in hepatocellular carcinoma patients after a hepatectomy.
Multiplication of alpha-fetoprotein and protein induced by vitamin K absence-II is a powerful predictor of prognosis and recurrence in hepatocellular carcinoma patients after a hepatectomy.
Hepatology research : the official journal of the Japan Society of Hepatology (2014-11-11)
Toshiya Kamiyama, Hideki Yokoo, Tatsuhiko Kakisaka, Tatsuya Orimo, Kenji Wakayama, Hirofumi Kamachi, Yosuke Tsuruga, Kenichiro Yamashita, Tsuyoshi Shimamura, Satoru Todo, Akinobu Taketomi
ABSTRACT
To evaluate the oncological implications of multiplication of α-fetoprotein (AFP) and protein induced by vitamin K absence or antagonists-II (PIVKA-II) in patients with hepatocellular carcinoma (HCC). Data were prospectively collected from 516 consecutive patients who underwent a curative primary hepatectomy for HCC between 1998 and 2010. The AP-factor (AFP × PIVKA-II) was evaluated in relation to 2-year survival outcomes by receiver-operator curve analysis to determine the cut-off values. Patient survival, recurrence-free survival and risk factors were analyzed in accordance with the preoperative AP-factor. The AP-factor was categorized into three groups depending on the serum concentrations of AFP and PIVKA-II as follows: AP1 (n = 206; AFP < 200 ng/mL and PIVKA-II < 100 mAU/mL), AP2 (n = 152; AFP × PIVKA-II < 10(5) ) and AP3 (n = 158; AFP × PIVKA-II ≥ 10(5) ). The AP-factor was found to be significantly related to pathological factors such as differentiation, portal vein invasion, hepatic vein invasion and intrahepatic metastasis. Multivariate analysis was performed to identify the risk factors for survival and recurrence. Albumin, AP-factor and pathological factors including portal vein invasion, hepatic vein invasion and intrahepatic metastasis are independent risk factors for survival. Tumor number, AP-factor, and a non-cancerous liver were determinants of recurrence. The AP-factor is closely related to differentiation and microscopic vascular invasion, and was selected by multivariate analysis as an independent factor for survival and recurrence, in HCC. Patients hopeful of obtaining good outcomes after a hepatectomy could be selected by the AP-factor evaluation.
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)