Merck
  • Telomere dysfunction reduces microglial numbers without fully inducing an aging phenotype.
Telomere dysfunction reduces microglial numbers without fully inducing an aging phenotype.
Neurobiology of aging (2015-04-22)
Asif Manzoor Khan, Alicia A Babcock, Hamid Saeed, Christa Løth Myhre, Moustapha Kassem, Bente Finsen
ABSTRACT
The susceptibility of the aging brain to neurodegenerative disease may in part be attributed to cellular aging of the microglial cells that survey it. We investigated the effect of cellular aging induced by telomere shortening on microglia by the use of mice lacking the telomerase RNA component (TERC) and design-based stereology. TERC knockout (KO) mice had a significantly reduced number of CD11b(+) microglia in the dentate gyrus. Because of an even greater reduction in dentate gyrus volume, microglial density was, however, increased. Microglia in TERC KO mice maintained a homogenous distribution and normal expression of CD45 and CD68 and the aging marker, ferritin, but were morphologically distinct from microglia in both adult and old wild-type mice. TERC KO mice also showed increased cellular apoptosis and impaired spatial learning. Our results suggest that individual microglia are relatively resistant to telomerase deficiency during steady state conditions, despite an overall reduction in microglial numbers. Furthermore, telomerase deficiency and aging may provide disparate cues leading to distinct changes in microglial morphology and phenotype.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd68
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprc
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%