Merck
  • Epigenetic alterations and decreasing insecticide sensitivity of the Asian tiger mosquito Aedes albopictus.
Epigenetic alterations and decreasing insecticide sensitivity of the Asian tiger mosquito Aedes albopictus.
Ecotoxicology and environmental safety (2015-07-21)
A Oppold, A Kreß, J Vanden Bussche, J B Diogo, U Kuch, J Oehlmann, M B Vandegehuchte, R Müller
ABSTRACT
A range of environmental factors, including chemicals, can affect epigenetic processes in organisms leading to variations in phenotype. Thus, epigenetics displays an important environmentally responsive element. The transgenerational impact of environmental stressors on DNA methylation and phenotype was the focus of this study. The influence of two known DNA methylation-changing agents, the phytoestrogen genistein and the fungicide vinclozolin, on the overall DNA methylation level in the Asian tiger mosquito Aedes albopictus was investigated. The experiment comprised four generations in a full life-cycle design with an exposed parental generation and three consecutive non-exposed offspring generations. Application of the methylation agents to the parental generation of the study led to an alteration of the global DNA methylation level of the exposed individuals and those in two subsequent generations. The phenotypic variability of the offspring generations was assessed by examining their insecticide sensitivity. Here, a significant decrease in sensitivity (p<0.01) towards the model insecticide imidacloprid revealed alterations of the mosquito's phenotype in two subsequent generations. Thus, the evaluation of A. albopictus from an epigenetic perspective can contribute important information to the study of the high adaptability of this invasive disease vector to new environments, and its underlying mechanisms.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)